Assay Details
| Gene Symbol: LINC00996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034033.1 | 2 - 3 | 790 | 104 | ||
| GenBank mRNA | AK097158.1 | - | 2 - 3 | 692 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00996| Gene Symbol: LINC00996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034033.1 | 2 - 3 | 790 | 104 | ||
| GenBank mRNA | AK097158.1 | - | 2 - 3 | 692 | 104 | |
Set Membership: |
|