Assay Details
Gene Symbol: HTR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310284.1 | - | 1 - 2 | 98 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000287907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | ENST00000486819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009796.hg.1 | NM_024012.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000430851.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009043.hg.1 | ENST00000434708.1 | Ensembl |
