Assay Details
Gene Symbol: LOC441242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103527.2 | 2 - 3 | 94 | 70 | ||
GenBank mRNA | BX647963.1 | - | 3 - 4 | 345 | 70 | |
CN483231.1 | - | 2 - 3 | 85 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
