Assay Details
| Gene Symbol: TCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206938.1 | NP_001193867.1 | 9 | 4364 | 118 | |
| NM_001206941.1 | NP_001193870.1 | 8 | 3709 | 118 | ||
| NM_014719.2 | NP_055534.1 | 9 | 4298 | 118 | ||
| XM_005250074.3 | XP_005250131.1 | 9 | 4338 | 118 | ||
| XM_005250075.2 | XP_005250132.1 | 9 | 4340 | 118 | ||
| XM_005250076.3 | XP_005250133.1 | 9 | 4375 | 118 | ||
| XM_005250077.2 | XP_005250134.1 | 10 | 4504 | 118 | ||
| XM_006716191.2 | XP_006716254.1 | 9 | 4304 | 118 | ||
| XM_006716192.3 | XP_006716255.1 | 9 | 4473 | 118 | ||
| XM_017012833.1 | XP_016868322.1 | 9 | 4396 | 118 | ||
| XM_017012834.1 | XP_016868323.1 | 9 | 4366 | 118 | ||
| XM_017012835.1 | XP_016868324.1 | 10 | 4530 | 118 | ||
| XM_017012836.1 | XP_016868325.1 | 9 | 4330 | 118 | ||
| GenBank mRNA | AF119896.2 | - | 1 | 118 | ||
| BC039457.1 | - | 1 | 258 | 118 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000355951.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000392900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000460532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000478172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000479870.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000485416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | ENST00000491908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_001206941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012898.hg.1 | NM_014719.2 | Refseq |

TCAF1
Genomic Map