Assay Details
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017012584.1 | XP_016868073.1 | 2 - 3 | 1152 | 90 | |
GenBank mRNA | AK301871.1 | - | 2 - 3 | 172 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK301871.1, XM_017012584.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
