Assay Details
Gene Symbol: LOC349160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF279773.1 | - | 2 - 3 | 278 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000599888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000599888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000425981.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000439694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000586239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000592183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000593789.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000597642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000598184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000599888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000599888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | ENST00000608269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012719.hg.1 | NR_046103.1 | Refseq |
