Assay Details
Gene Symbol: EFCAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027068.1 | 2 - 3 | 342 | 90 | ||
GenBank mRNA | BC105284.1 | - | 2 - 3 | 317 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
