Assay Details
| Gene Symbol: KDM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030647.1 | NP_085150.1 | 15 - 16 | 2020 | 70 | |
| XM_011516587.2 | XP_011514889.1 | 9 - 10 | 1045 | 70 | ||
| GenBank mRNA | AB051505.1 | - | 3 - 4 | 325 | 70 | |
| AK131497.1 | - | 15 - 16 | 2021 | 70 | ||
| BX641017.1 | - | 7 - 8 | 800 | 70 | ||
| BX649115.1 | - | 10 - 11 | 1287 | 70 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
midbrain development
histone H3-K9 demethylation
histone H4-K20 demethylation
positive regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
histone H3-K27 demethylation
iron ion binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
histone demethylase activity
histone demethylase activity (H3-K9 specific)
methylated histone binding
histone demethylase activity (H4-K20 specific)
histone demethylase activity (H3-K36 specific)
histone demethylase activity (H3-K27 specific)
transcription, DNA-templated
midbrain development
histone H3-K9 demethylation
histone H4-K20 demethylation
positive regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
histone H3-K27 demethylation
iron ion binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
histone demethylase activity
histone demethylase activity (H3-K9 specific)
methylated histone binding
histone demethylase activity (H4-K20 specific)
histone demethylase activity (H3-K36 specific)
histone demethylase activity (H3-K27 specific)
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000397560.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000472616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | ENST00000478996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012798.hg.1 | NM_030647.1 | Refseq |

KDM7A
Genomic Map