Assay Details
| Gene Symbol: LINC-PINT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034120.1 | 2 - 3 | 404 | 73 | ||
| NR_110472.1 | 2 - 3 | 404 | 73 | |||
| NR_110473.1 | 2 - 3 | 404 | 73 | |||
| GenBank mRNA | AK095549.1 | - | 2 - 3 | 407 | 73 | |
| GU321462.1 | - | 2 - 3 | 225 | 73 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000385015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_029503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_029517.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000385015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_029503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_029517.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |

LINC-PINT
Genomic Map