Assay Details
Gene Symbol: GPR141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC043356.1 | - | 3 - 4 | 468 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000334425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000334425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | NM_181791.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013354.hg.1 | NM_181791.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000334425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000334425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000447769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000450180.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000461610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | ENST00000467202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | NM_181791.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013354.hg.1 | NM_181791.2 | Refseq |
