Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |