Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000525030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006694.hg.1 |
ENST00000607111.2 |
Ensembl |