Assay Details
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290076.1 | NP_001277005.1 | 2 - 3 | 394 | 103 | |
GenBank mRNA | AK302459.1 | - | 2 - 3 | 394 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK302459.1, NM_001290076.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
