Assay Details
| Gene Symbol: TMEM178B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195278.1 | NP_001182207.1 | 2 - 3 | 576 | 88 | |
| XM_011515705.2 | XP_011514007.1 | 2 - 3 | 1406 | 88 | ||
| XM_017011636.1 | XP_016867125.1 | 2 - 3 | 1400 | 88 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000563442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000565468.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | ENST00000610315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009386.hg.1 | NM_001195278.1 | Refseq |

TMEM178B
Genomic Map