Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
ENST00000395145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011503.hg.1 |
NM_001306.3 |
Refseq |