Assay Details
| Gene Symbol: KIAA0196-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DA611863.1 | - | 1 - 2 | 96 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0196-AS1| Gene Symbol: KIAA0196-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DA611863.1 | - | 1 - 2 | 96 | 82 | |
Set Membership: |
|