| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000519242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522302.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
ENST00000522807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010878.hg.1 |
NR_125389.1 |
Refseq |