Assay Details
Gene Symbol: LSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014462.2 | NP_055277.1 | 3 - 4 | 639 | 108 | |
NR_045493.1 | 3 - 4 | 361 | 108 | |||
GenBank mRNA | AF000177.1 | - | 3 - 4 | 393 | 108 | |
AF147348.1 | - | 3 - 4 | 282 | 108 | ||
AJ238094.1 | - | 3 - 4 | 417 | 108 | ||
AK312159.1 | - | 3 - 4 | 392 | 108 | ||
BC001767.1 | - | 3 - 4 | 326 | 3353833 | 108 | |
BE670269.1 | - | 1 - 2 | 108 | |||
BU941962.1 | - | 3 - 4 | 364 | 108 | ||
KU178606.1 | - | 3 - 4 | 229 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
deadenylation-dependent decapping of nuclear-transcribed mRNA
RNA splicing, via transesterification reactions
mRNA processing
RNA splicing
stem cell population maintenance
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
mRNA binding
protein binding
pre-mRNA binding
poly(A) RNA binding
deadenylation-dependent decapping of nuclear-transcribed mRNA
RNA splicing, via transesterification reactions
mRNA processing
RNA splicing
stem cell population maintenance
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
mRNA binding
protein binding
pre-mRNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007324.hg.1 | ENST00000520598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007324.hg.1 | ENST00000520598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007324.hg.1 | ENST00000520598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007324.hg.1 | ENST00000520598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
