Assay Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC046930.1 | - | 1 | 1920 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
