Assay Details
| Gene Symbol: KIAA0196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014846.3 | NP_055661.3 | 2 - 3 | 513 | 98 | |
| XM_011517409.1 | XP_011515711.1 | 2 - 3 | 490 | 98 | ||
| XM_017014113.1 | XP_016869602.1 | 2 - 3 | 718 | 98 | ||
| GenBank mRNA | AK223628.1 | - | 2 - 3 | 461 | 98 | |
| AK291032.1 | - | 2 - 3 | 336 | 98 | ||
| BC026951.1 | - | 1 - 2 | 264 | 3882443 | 98 | |
| BC106015.1 | - | 2 - 3 | 402 | 6059577 | 98 | |
| D83780.1 | - | 2 - 3 | 457 | 98 | ||
More Information
Set Membership: |
|

KIAA0196
Genomic Map