Assay Details
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006716500.2 | XP_006716563.1 | 1 - 2 | 94 | 110 | |
XM_011516805.1 | XP_011515107.1 | 1 - 2 | 91 | 110 | ||
XM_017012998.1 | XP_016868487.1 | 1 - 2 | 94 | 110 | ||
XM_017012999.1 | XP_016868488.1 | 1 - 2 | 94 | 110 | ||
GenBank mRNA | AK311257.1 | - | 1 - 2 | 66 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716500.2, XM_011516805.1, XM_017012998.1, XM_017012999.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
Notch signaling pathway
peptidyl-tyrosine dephosphorylation
positive regulation of vascular permeability
endothelial cell migration
regulation of vascular endothelial growth factor signaling pathway
prenylated protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
Notch signaling pathway
peptidyl-tyrosine dephosphorylation
positive regulation of vascular permeability
endothelial cell migration
regulation of vascular endothelial growth factor signaling pathway
prenylated protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000329397.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000349124.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000520105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000521578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000523147.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | ENST00000524028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_007079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009113.hg.1 | NM_032611.2 | Refseq |
