| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |