Assay Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080416.3 | NP_001073885.1 | 1 | 177 | 62 | |
NM_001144755.2 | NP_001138227.1 | 1 | 177 | 62 | ||
NM_001294282.1 | NP_001281211.1 | 1 | 177 | 62 | ||
XM_011517533.2 | XP_011515835.1 | 1 | 218 | 62 | ||
XM_011517535.2 | XP_011515837.1 | 1 | 218 | 62 | ||
GenBank mRNA | BC032233.1 | - | 1 | 120 | 62 | |
BC046930.1 | - | 1 | 1175 | 62 | ||
CN482561.1 | - | 1 | 173 | 62 | ||
DR003430.1 | - | 1 | 185 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001080416.3, NM_001144755.2, NM_001294282.1, XM_011517533.2, XM_011517535.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
