Assay Details
| Gene Symbol: PPP1R16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272350.2 | XP_005272407.1 | 12 | 2728 | 89 | |
| XM_011517349.2 | XP_011515651.1 | 11 | 1579 | 89 | ||
| XM_017013912.1 | XP_016869401.1 | 13 | 2797 | 89 | ||
| XM_017013913.1 | XP_016869402.1 | 12 | 2761 | 89 | ||
| XM_017013914.1 | XP_016869403.1 | 13 | 2835 | 89 | ||
| XM_017013915.1 | XP_016869404.1 | 12 | 2792 | 89 | ||
| GenBank mRNA | AK090471.1 | - | 9 | 4094 | 89 | |
| BC007854.2 | - | 10 | 1802 | 4298333 | 89 | |
| BC049841.1 | - | 10 | 2054 | 4991947 | 89 | |
| BC053506.1 | - | 10 | 2143 | 2823232 | 89 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |

PPP1R16A
Genomic Map