Assay Details
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006716749.3 | XP_006716812.1 | 21 | 6127 | 80 | |
XM_011517836.2 | XP_011516138.1 | 21 | 6263 | 80 | ||
XM_011517837.2 | XP_011516139.1 | 21 | 6260 | 80 | ||
GenBank mRNA | AK058117.1 | - | 1 | 1325 | 80 | |
BC024252.1 | - | 1 | 1352 | 80 | ||
BC047488.1 | - | 1 | 1348 | 80 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011517881.2 | XP_011516183.1 | 21 | 5819 | 80 | |
XM_011517883.2 | XP_011516185.1 | 20 | 5765 | 80 | ||
XM_011517885.2 | XP_011516187.1 | 15 | 4910 | 80 | ||
XM_017014728.1 | XP_016870217.1 | 15 | 4907 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
