Assay Details
| Gene Symbol: LOC730098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320037.1 | NP_001306966.1 | 2 - 3 | 143 | 111 | |
| NM_001320038.1 | NP_001306967.1 | 1 - 2 | 440 | 111 | ||
| NR_135148.1 | 2 - 3 | 143 | 111 | |||
| XM_017015106.1 | XP_016870595.1 | 2 - 3 | 300 | 111 | ||
| GenBank mRNA | AK296005.1 | - | 1 - 2 | 400 | 111 | |
| BC066358.1 | - | 1 - 2 | 433 | 111 | ||
| BM695963.1 | - | 2 - 3 | 112 | 111 | ||
| HY158194.1 | - | 2 - 3 | 138 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296005.1, NM_001320037.1, NM_001320038.1, XM_017015106.1 transcripts.
Set Membership: |
|

LOC730098
Genomic Map