Assay Details
| Gene Symbol: LOC730098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320037.1 | NP_001306966.1 | 3 - 4 | 205 | 71 | |
| NM_001320038.1 | NP_001306967.1 | 2 - 3 | 502 | 71 | ||
| XM_017015106.1 | XP_016870595.1 | 3 - 4 | 362 | 71 | ||
| GenBank mRNA | AK296005.1 | - | 2 - 3 | 462 | 71 | |
| BC066358.1 | - | 2 - 3 | 495 | 71 | ||
| HY158194.1 | - | 3 - 4 | 200 | 71 | ||
More Information
Set Membership: |
|

LOC730098
Genomic Map