Assay Details
| Gene Symbol: ST6GALNAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518610.2 | XP_011516912.1 | 2 - 3 | 886 | 92 | |
| XM_017014659.1 | XP_016870148.1 | 2 - 3 | 886 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518610.2, XM_017014659.1 transcripts.
Set Membership: |
|

ST6GALNAC6
Genomic Map