Assay Details
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136024.3 | NP_001129496.1 | 5 - 6 | 850 | 89 | |
NM_001291867.1 | NP_001278796.1 | 5 - 6 | 1445 | 89 | ||
NM_001291868.1 | NP_001278797.1 | 4 - 5 | 787 | 89 | ||
NM_198270.3 | NP_938011.1 | 4 - 5 | 1382 | 89 | ||
XM_011545528.2 | XP_011543830.1 | 2 - 3 | 994 | 89 | ||
GenBank mRNA | AK302437.1 | - | 5 - 6 | 1631 | 89 | |
AY436752.1 | - | 4 - 5 | 1382 | 89 | ||
AY456992.1 | - | 5 - 6 | 850 | 89 | ||
AY456993.1 | - | 4 - 5 | 1505 | 89 | ||
BC136415.1 | - | 5 - 6 | 1171 | 9020403 | 89 | |
BC171763.1 | - | 5 - 6 | 1171 | 9054416 | 89 | |
CR936788.1 | - | 4 - 5 | 556 | 89 | ||
GQ988776.1 | - | 5 - 6 | 1110 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000380060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000398097.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000485305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000615422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | ENST00000617601.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001136024.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291867.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_001291868.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006709.hg.1 | NM_198270.3 | Refseq |
