Assay Details
| Gene Symbol: CASK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA961433.1 | - | 1 - 2 | 334 | 92 | |
| BE463818.1 | - | 1 - 2 | 334 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASK-AS1| Gene Symbol: CASK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA961433.1 | - | 1 - 2 | 334 | 92 | |
| BE463818.1 | - | 1 - 2 | 334 | 92 | ||
Set Membership: |
|