Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
ENST00000276055.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007112.hg.1 |
NM_019886.3 |
Refseq |