Assay Details
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 2956 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000376771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000418627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000476141.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000481090.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | ENST00000494495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007194.hg.1 | NM_002536.3 | Refseq |
