Assay Details
Gene Symbol: TIMM17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167947.2 | NP_001161419.1 | 4 - 5 | 425 | 72 | |
GenBank mRNA | CB993524.1 | - | 3 - 4 | 316 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000464663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000464663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000376582.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000396779.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000464663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000464663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000465150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000466995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000472645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000490755.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | ENST00000495490.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_001167947.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009634.hg.1 | NM_005834.4 | Refseq |
