Assay Details
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_072995.1 | 1 - 2 | 276 | 132 | ||
XM_017018848.1 | XP_016874337.1 | 1 - 2 | 85 | 132 | ||
GenBank mRNA | BX640916.1 | - | 1 - 2 | 245 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017018848.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000320460.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000378692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000547164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | ENST00000550916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_001270396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NM_152436.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008223.hg.1 | NR_072995.1 | Refseq |
