Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000499685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000501008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008425.hg.1 |
ENST00000618125.1 |
Ensembl |