Assay Details
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308340.1 | NP_001295269.1 | 2 - 3 | 508 | 121 | |
GenBank mRNA | AL712776.1 | - | 2 - 3 | 501 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000308742.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000548240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000551464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | ENST00000553138.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001013620.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007326.hg.1 | NM_001308340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000266483.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000538927.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | ENST00000541875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007295.hg.1 | NM_032834.3 | Refseq |
