Assay Details
Gene Symbol: LINC00507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046392.1 | 4 - 5 | 552 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009515.hg.1 | ENST00000521218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000540882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000542089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | ENST00000544645.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009356.hg.1 | NR_046392.1 | Refseq |
