Assay Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011534812.2 | XP_011533114.1 | 2 - 3 | 232 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011534812.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
