Assay Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005266181.3 | XP_005266238.1 | 2 - 3 | 618 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008653.hg.1 | ENST00000466506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008653.hg.1 | ENST00000466506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000434748.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000536075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000537804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000539264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000542306.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | ENST00000543453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009489.hg.1 | NM_001142641.1 | Refseq |
