Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000425031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
ENST00000444263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006488.hg.1 |
NR_046502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000420327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007813.hg.1 |
ENST00000435867.1 |
Ensembl |