Assay Details
Gene Symbol: LOC101927653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DB024932.1 | - | 2 - 3 | 303 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000550678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | ENST00000552201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011000.hg.1 | NR_120452.1 | Refseq |
