Assay Details
| Gene Symbol: LOC102724264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134483.1 | 1 - 2 | 318 | 75 | ||
| GenBank mRNA | AA868454.1 | - | 1 - 2 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724264| Gene Symbol: LOC102724264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134483.1 | 1 - 2 | 318 | 75 | ||
| GenBank mRNA | AA868454.1 | - | 1 - 2 | 75 | ||
Set Membership: |
|