Assay Details
| Gene Symbol: LOC101928834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120646.1 | 2 - 3 | 241 | 95 | ||
| GenBank mRNA | BC014575.1 | - | 2 - 3 | 245 | 95 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |

LOC101928834
Genomic Map