Assay Details
| Gene Symbol: ZNF33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130951.1 | 3 - 4 | 556 | 113 | ||
| GenBank mRNA | BC042873.1 | - | 3 - 4 | 1278 | 113 | |
| DA826003.1 | - | 3 - 4 | 283 | 113 | ||
More Information
Set Membership: |
|

ZNF33B
Genomic Map