Assay Details
Gene Symbol: PPIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540508.1 | XP_011538810.1 | 2 - 3 | 152 | 99 | |
XM_017000058.1 | XP_016855547.1 | 2 - 3 | 806 | 99 | ||
XM_017000059.1 | XP_016855548.1 | 2 - 3 | 152 | 99 | ||
GenBank mRNA | AK309438.1 | - | 2 - 3 | 148 | 99 | |
AK310052.1 | - | 2 - 3 | 152 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540508.1, XM_017000058.1, XM_017000059.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000304979.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000372550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000436387.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000440068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | ENST00000461351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008023.hg.1 | NM_006347.3 | Refseq |
