| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000374555.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
ENST00000612756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018197.hg.1 |
NR_027042.1 |
Refseq |