Assay Details
| Gene Symbol: AGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541236.2 | XP_011539538.1 | 19 | 9813 | 106 | |
| XM_017001016.1 | XP_016856505.1 | 19 | 9861 | 106 | ||
| XM_017001017.1 | XP_016856506.1 | 19 | 9852 | 106 | ||
| XM_017001018.1 | XP_016856507.1 | 16 | 9336 | 106 | ||
| XM_017001019.1 | XP_016856508.1 | 19 | 9901 | 106 | ||
| GenBank mRNA | AK125879.1 | - | 1 | 823 | 106 | |
| Gene Symbol: AGO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270575.3 | XP_005270632.1 | 1 | 1452 | 106 | |
| XM_017000523.1 | XP_016856012.1 | 1 | 1452 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005270575.3, XM_017000523.1 transcripts.
Set Membership: |
|

AGO1
Genomic Map