Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |