Assay Details
| Gene Symbol: PPP5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204284.1 | NP_001191213.1 | 7 - 8 | 1083 | 70 | |
| NM_006247.3 | NP_006238.1 | 8 - 9 | 1149 | 70 | ||
| XM_017026935.1 | XP_016882424.1 | 8 - 9 | 1144 | 70 | ||
| GenBank mRNA | AB209615.1 | - | 3 - 4 | 2626 | 70 | |
| AK094241.1 | - | 3 - 4 | 1355 | 70 | ||
| AK223222.1 | - | 8 - 9 | 1056 | 70 | ||
| AK293402.1 | - | 9 - 10 | 1007 | 70 | ||
| AK293475.1 | - | 4 - 5 | 664 | 70 | ||
| AK297095.1 | - | 9 - 10 | 995 | 70 | ||
| AK312683.1 | - | 8 - 9 | 1056 | 70 | ||
| BC000750.2 | - | 8 - 9 | 1042 | 70 | ||
| BC001831.2 | - | 8 - 9 | 1042 | 70 | ||
| BC001970.1 | - | 8 - 9 | 1106 | 3459309 | 70 | |
| BT007275.1 | - | 8 - 9 | 1046 | 70 | ||
| U25174.1 | - | 8 - 9 | 1024 | 70 | ||
| X89416.1 | - | 8 - 9 | 1029 | 70 | ||
More Information
Set Membership: |
|

PPP5C
Genomic Map