Assay Details
| Gene Symbol: ZNF571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX537401.1 | - | 5 | 3946 | 113 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000328550.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000358744.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000451802.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000590751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | ENST00000593133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_001290314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010574.hg.1 | NM_016536.4 | Refseq |

ZNF571
Genomic Map